Assay Details
| Gene Symbol: Sri | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007646778.1 | XP_007644968.1 | 5 - 6 | 561 | 64 | |
| GenBank mRNA | JP051217.1 | - | 5 - 6 | 561 | 64 | |
More Information
Set Membership: |
|

Sri
Genomic Map