Assay Details
| Gene Symbol: Yrdc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007642858.1 | XP_007641048.1 | 3 - 4 | 364 | 61 | |
| GenBank mRNA | JI869235.1 | - | 2 - 3 | 237 | 61 | |
More Information
Set Membership: |
|

Yrdc
Genomic Map