Assay Details
| Gene Symbol: Nmnat3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007641472.1 | XP_007639662.1 | 3 - 4 | 501 | 68 | |
| XM_007641477.1 | XP_007639667.1 | 4 - 5 | 569 | 68 | ||
| XM_007641481.1 | XP_007639671.1 | 3 - 4 | 533 | 68 | ||
| XM_007641484.1 | XP_007639674.1 | 3 - 4 | 502 | 68 | ||
| GenBank mRNA | JP044191.1 | - | 2 - 3 | 249 | 68 | |
More Information
Set Membership: |
|

Nmnat3
Genomic Map