Assay Details
| Gene Symbol: LOC100750533 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003496336.2 | XP_003496384.1 | 3 - 4 | 560 | 90 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100750533| Gene Symbol: LOC100750533 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003496336.2 | XP_003496384.1 | 3 - 4 | 560 | 90 | |