Assay Details
| Gene Symbol: Acyp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003502768.2 | XP_003502816.1 | 2 - 3 | 133 | 103 | |
| XM_007643386.1 | XP_007641576.1 | 2 - 3 | 385 | 103 | ||
| XM_007643387.1 | XP_007641577.1 | 2 - 3 | 213 | 103 | ||
| GenBank mRNA | JI876095.1 | - | 1 - 2 | 90 | 103 | |
More Information
Set Membership: |
|

Acyp1
Genomic Map