Assay Details
| Gene Symbol: Med28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003505976.2 | XP_003506024.1 | 1 - 2 | 180 | 59 | |
| GenBank mRNA | JI866635.1 | - | 3 - 4 | 59 | ||
| JP053201.1 | - | 1 - 2 | 163 | 59 | ||
More Information
Set Membership: |
|

Med28
Genomic Map