Assay Details
| Gene Symbol: Cdip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003506346.2 | XP_003506394.1 | 5 - 6 | 776 | 60 | |
| XM_007646857.1 | XP_007645047.1 | 5 - 6 | 684 | 60 | ||
| XM_007646858.1 | XP_007645048.1 | 5 - 6 | 672 | 60 | ||
| GenBank mRNA | JI888226.1 | - | 5 - 6 | 638 | 60 | |
| JP062851.1 | - | 5 - 6 | 638 | 60 | ||
More Information
Set Membership: |
|

Cdip1
Genomic Map