Assay Details
| Gene Symbol: Gpr137b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003507584.2 | XP_003507632.1 | 3 - 4 | 668 | 70 | |
| GenBank mRNA | JI868302.1 | - | 1 - 2 | 223 | 70 | |
| JP044702.1 | - | 1 - 2 | 223 | 70 | ||
More Information
Set Membership: |
|

Gpr137b
Genomic Map