Assay Details
| Gene Symbol: Fam196a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003510496.1 | XP_003510544.1 | 2 - 3 | 1157 | 60 | |
| XM_007651120.1 | XP_007649310.1 | 2 - 3 | 1157 | 60 | ||
More Information
Set Membership: |
|

Fam196a
Genomic Map