Assay Details
| Gene Symbol: Tor1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003513974.2 | XP_003514022.1 | 2 - 3 | 501 | 61 | |
| GenBank mRNA | JI872315.1 | - | 2 - 3 | 375 | 61 | |
| JP059117.1 | - | 2 - 3 | 375 | 61 | ||
More Information
Set Membership: |
|

Tor1a
Genomic Map