Assay Details
| Gene Symbol: Rela | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007653319.1 | XP_007651509.1 | 3 - 4 | 418 | 60 | |
| GenBank mRNA | JI863893.1 | - | 3 - 4 | 419 | 60 | |
| JP060285.1 | - | 3 - 4 | 418 | 60 | ||
More Information
Set Membership: |
|

Rela
Genomic Map