Assay Details
| Gene Symbol: Eif3h | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007619178.1 | XP_007617368.1 | 3 - 4 | 277 | 60 | |
| GenBank mRNA | JP055479.1 | - | 1 - 2 | 285 | 60 | |
More Information
Set Membership: |
|

Eif3h
Genomic Map