Assay Details
| Gene Symbol: Pip4k2c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007648113.1 | XP_007646303.1 | 8 - 9 | 1090 | 67 | |
| GenBank mRNA | JI883957.1 | - | 2 - 3 | 347 | 67 | |
| JP065691.1 | - | 2 - 3 | 347 | 67 | ||
More Information
Set Membership: |
|

Pip4k2c
Genomic Map