Assay Details
| Gene Symbol: Il17f | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007649727.1 | XP_007647917.1 | 2 - 3 | 235 | 58 | |
| XM_007649729.1 | XP_007647919.1 | 3 - 4 | 251 | 58 | ||
More Information
Set Membership: |
|

Il17f
Genomic Map