Assay Details
| Gene Symbol: Traf6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003497459.2 | XP_003497507.1 | 6 - 7 | 942 | 70 | |
| GenBank mRNA | JI893980.1 | - | 5 - 6 | 634 | 70 | |
More Information
Set Membership: |
|

Traf6
Genomic Map