Assay Details
| Gene Symbol: Hyal2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003500400.2 | XP_003500448.1 | 2 - 3 | 1225 | 76 | |
| GenBank mRNA | JI885550.1 | - | 1 - 2 | 529 | 76 | |
More Information
Set Membership: |
|

Hyal2
Genomic Map