Assay Details
| Gene Symbol: Tnfsf14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003501773.1 | XP_003501821.1 | 2 - 3 | 252 | 77 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tnfsf14| Gene Symbol: Tnfsf14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003501773.1 | XP_003501821.1 | 2 - 3 | 252 | 77 | |
Set Membership: |
|