Assay Details
| Gene Symbol: Exog | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003505922.2 | XP_003505970.1 | 3 - 4 | 460 | 74 | |
| GenBank mRNA | JI896513.1 | - | 2 - 3 | 297 | 74 | |
| JP037634.1 | - | 2 - 3 | 297 | 74 | ||
More Information
Set Membership: |
|

Exog
Genomic Map