Assay Details
| Gene Symbol: Pla2g2e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003506895.2 | XP_003506943.1 | 2 - 3 | 293 | 71 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pla2g2e| Gene Symbol: Pla2g2e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003506895.2 | XP_003506943.1 | 2 - 3 | 293 | 71 | |
Set Membership: |
|