Assay Details
| Gene Symbol: Gnpnat1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003510726.2 | XP_003510774.1 | 3 - 4 | 335 | 68 | |
| XM_007651320.1 | XP_007649510.1 | 3 - 4 | 257 | 68 | ||
| XM_007651321.1 | XP_007649511.1 | 3 - 4 | 340 | 68 | ||
| GenBank mRNA | JI896787.1 | - | 3 - 4 | 304 | 68 | |
| JP049865.1 | - | 3 - 4 | 304 | 68 | ||
More Information
Set Membership: |
|

Gnpnat1
Genomic Map