Assay Details
| Gene Symbol: Gpr125 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003511951.2 | XP_003511999.1 | 18 - 19 | 2549 | 95 | |
| GenBank mRNA | JI885131.1 | - | 4 - 5 | 661 | 95 | |
| JP043372.1 | - | 4 - 5 | 661 | 95 | ||
More Information
Set Membership: |
|

Gpr125
Genomic Map