Assay Details
Gene Symbol: Ddx19b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003513682.2 | XP_003513730.1 | 6 - 7 | 590 | 62 | |
GenBank mRNA | JI874764.1 | - | 6 - 7 | 595 | 62 | |
JP055977.1 | - | 6 - 7 | 594 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
