Assay Details
| Gene Symbol: Sdf4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007650625.1 | XP_007648815.1 | 2 - 3 | 500 | 73 | |
| XM_007650626.1 | XP_007648816.1 | 2 - 3 | 553 | 73 | ||
| GenBank mRNA | JI875479.1 | - | 1 - 2 | 325 | 73 | |
More Information
Set Membership: |
|

Sdf4
Genomic Map