Assay Details
| Gene Symbol: Pvrl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007645963.1 | XP_007644153.1 | 4 - 5 | 815 | 78 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pvrl2| Gene Symbol: Pvrl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007645963.1 | XP_007644153.1 | 4 - 5 | 815 | 78 | |