Assay Details
| Gene Symbol: Tmeff1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007644550.1 | XP_007642740.1 | 7 - 8 | 622 | 73 | |
| GenBank mRNA | JI881566.1 | - | 2 - 3 | 74 | ||
More Information
Set Membership: |
|

Tmeff1
Genomic Map