Assay Details
| Gene Symbol: LOC100767524 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003514408.2 | XP_003514456.1 | 4 - 5 | 530 | 126 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100767524| Gene Symbol: LOC100767524 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003514408.2 | XP_003514456.1 | 4 - 5 | 530 | 126 | |
Set Membership: |
|