Assay Details
| Gene Symbol: Rbbp7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007649847.1 | XP_007648037.1 | 1 - 2 | 463 | 74 | |
| GenBank mRNA | JI895546.1 | - | 1 - 2 | 463 | 74 | |
More Information
Set Membership: |
|

Rbbp7
Genomic Map