Assay Details
| Gene Symbol: Capns1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007646060.1 | XP_007644250.1 | 7 - 8 | 468 | 69 | |
| GenBank mRNA | JI889275.1 | - | 6 - 7 | 622 | 69 | |
More Information
Set Membership: |
|

Capns1
Genomic Map