Assay Details
| Gene Symbol: Dnaja1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007651996.1 | XP_007650186.1 | 4 - 5 | 759 | 81 | |
| GenBank mRNA | AB104412.1 | - | 1 | 759 | 81 | |
| JP051891.1 | - | 4 - 5 | 912 | 81 | ||
More Information
Set Membership: |
|

Dnaja1
Genomic Map