Assay Details
| Gene Symbol: Lrrc74 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007640408.1 | XP_007638598.1 | 2 - 3 | 134 | 101 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lrrc74| Gene Symbol: Lrrc74 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007640408.1 | XP_007638598.1 | 2 - 3 | 134 | 101 | |
Set Membership: |
|