Assay Details
| Gene Symbol: Zswim1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003500537.2 | XP_003500585.1 | 3 | 1203 | 72 | |
| XM_007641306.1 | XP_007639496.1 | 1 | 723 | 72 | ||
| GenBank mRNA | JP057557.1 | - | 2 | 910 | 72 | |
More Information
Set Membership: |
|

Zswim1
Genomic Map