Assay Details
| Gene Symbol: Dyx1c1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003495027.2 | XP_003495075.1 | 7 - 8 | 1776 | 81 | |
| XM_003495028.2 | XP_003495076.1 | 7 - 8 | 1776 | 81 | ||
More Information
Set Membership: |
|

Dyx1c1
Genomic Map