Assay Details
| Gene Symbol: Dyx1c1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003495027.2 | XP_003495075.1 | 8 - 9 | 1939 | 60 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dyx1c1| Gene Symbol: Dyx1c1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003495027.2 | XP_003495075.1 | 8 - 9 | 1939 | 60 | |
Set Membership: |
|