Assay Details
| Gene Symbol: Epb42 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003498079.1 | XP_003498127.1 | 9 - 10 | 1612 | 60 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Epb42| Gene Symbol: Epb42 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003498079.1 | XP_003498127.1 | 9 - 10 | 1612 | 60 | |
Set Membership: |
|