Assay Details
| Gene Symbol: Nuak2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003501202.2 | XP_003501250.1 | 3 - 4 | 618 | 57 | |
| XM_007641898.1 | XP_007640088.1 | 3 - 4 | 503 | 57 | ||
More Information
Set Membership: |
|

Nuak2
Genomic Map