Assay Details
| Gene Symbol: Smim19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003503309.2 | XP_003503357.1 | 3 - 4 | 969 | 76 | |
| XM_007643907.1 | XP_007642097.1 | 3 - 4 | 394 | 76 | ||
| XM_007643908.1 | XP_007642098.1 | 3 - 4 | 1791 | 76 | ||
| XM_007643909.1 | XP_007642099.1 | 3 - 4 | 333 | 76 | ||
| GenBank mRNA | JI862721.1 | - | 2 - 3 | 260 | 76 | |
| JP037173.1 | - | 3 - 4 | 336 | 76 | ||
More Information
Set Membership: |
|

Smim19
Genomic Map