Assay Details
| Gene Symbol: Tcp10l2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003504054.2 | XP_003504102.1 | 4 - 5 | 644 | 87 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tcp10l2| Gene Symbol: Tcp10l2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003504054.2 | XP_003504102.1 | 4 - 5 | 644 | 87 | |
Set Membership: |
|