Assay Details
| Gene Symbol: Gorab | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003505527.2 | XP_003505575.1 | 2 - 3 | 475 | 75 | |
| XM_007646025.1 | XP_007644215.1 | 2 - 3 | 478 | 75 | ||
| GenBank mRNA | JI882007.1 | - | 2 - 3 | 481 | 75 | |
| JP057432.1 | - | 2 - 3 | 481 | 75 | ||
More Information
Set Membership: |
|

Gorab
Genomic Map