Assay Details
| Gene Symbol: Zfand6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003514120.2 | XP_003514168.1 | 3 - 4 | 386 | 65 | |
| XM_003514121.2 | XP_003514169.1 | 2 - 3 | 243 | 65 | ||
| XM_007654313.1 | XP_007652503.1 | 2 - 3 | 323 | 65 | ||
| XM_007654314.1 | XP_007652504.1 | 2 - 3 | 243 | 65 | ||
| GenBank mRNA | JI869636.1 | - | 1 - 2 | 172 | 65 | |
| JP040932.1 | - | 2 - 3 | 298 | 65 | ||
More Information
Set Membership: |
|

Zfand6
Genomic Map