Assay Details
Gene Symbol: Fut7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003514669.1 | XP_003514717.1 | 1 - 2 | 167 | 117 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
