Assay Details
| Gene Symbol: Zkscan7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007641419.1 | XP_007639609.1 | 3 - 4 | 694 | 70 | |
| XM_007641420.1 | XP_007639610.1 | 3 - 4 | 694 | 70 | ||
More Information
Set Membership: |
|

Zkscan7
Genomic Map