Assay Details
Gene Symbol: Dct | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003495235.1 | XP_003495283.1 | 5 - 6 | 1042 | 61 | |
XM_007623836.1 | XP_007622026.1 | 5 - 6 | 1042 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
