Assay Details
| Gene Symbol: Lgalsl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003498108.2 | XP_003498156.1 | 2 - 3 | 162 | 65 | |
| GenBank mRNA | JI879323.1 | - | 1 - 2 | 100 | 65 | |
More Information
Set Membership: |
|

Lgalsl
Genomic Map