Assay Details
| Gene Symbol: Adamtsl5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003502572.2 | XP_003502620.1 | 10 - 11 | 1321 | 60 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Adamtsl5| Gene Symbol: Adamtsl5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003502572.2 | XP_003502620.1 | 10 - 11 | 1321 | 60 | |
Set Membership: |
|