Assay Details
| Gene Symbol: Rab23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003507909.2 | XP_003507957.1 | 3 - 4 | 449 | 77 | |
| XM_007648492.1 | XP_007646682.1 | 3 - 4 | 451 | 77 | ||
| XM_007648493.1 | XP_007646683.1 | 3 - 4 | 429 | 77 | ||
| XM_007648494.1 | XP_007646684.1 | 3 - 4 | 470 | 77 | ||
| XM_007648495.1 | XP_007646685.1 | 3 - 4 | 423 | 77 | ||
| XM_007648496.1 | XP_007646686.1 | 3 - 4 | 445 | 77 | ||
More Information
Set Membership: |
|

Rab23
Genomic Map