Assay Details
| Gene Symbol: Carkd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003509242.2 | XP_003509290.2 | 6 - 7 | 607 | 70 | |
| GenBank mRNA | JI895127.1 | - | 2 - 3 | 157 | 70 | |
More Information
Set Membership: |
|

Carkd
Genomic Map