Assay Details
| Gene Symbol: Carkd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003509242.2 | XP_003509290.2 | 7 - 8 | 731 | 69 | |
| GenBank mRNA | JI895127.1 | - | 3 - 4 | 281 | 69 | |
More Information
Set Membership: |
|

Carkd
Genomic Map