Assay Details
| Gene Symbol: Cst11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003509870.1 | XP_003509918.1 | 2 - 3 | 328 | 79 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cst11| Gene Symbol: Cst11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003509870.1 | XP_003509918.1 | 2 - 3 | 328 | 79 | |
Set Membership: |
|