Assay Details
| Gene Symbol: Nabp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003512328.2 | XP_003512376.1 | 4 - 5 | 820 | 78 | |
| GenBank mRNA | JI893190.1 | - | 3 - 4 | 255 | 78 | |
| JP050285.1 | - | 3 - 4 | 255 | 78 | ||
More Information
Set Membership: |
|

Nabp1
Genomic Map