Assay Details
| Gene Symbol: Mat2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007641637.1 | XP_007639827.1 | 3 - 4 | 622 | 77 | |
| GenBank mRNA | JI877952.1 | - | 3 - 4 | 622 | 77 | |
More Information
Set Membership: |
|

Mat2b
Genomic Map